Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006089.hg.1 |
ENST00000268459.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006089.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006089.hg.1 |
TC16000440.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006088.hg.1 |
ENST00000268459.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006088.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006088.hg.1 |
TC16000440.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600156949.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156949.hg.1 |
ENST00000564336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156949.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600156949.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156949.hg.1 |
ENST00000564336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156949.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600156949.hg.1 |
TC1600007819.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600156946.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156946.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600156946.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156946.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600156946.hg.1 |
TC1600007819.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600156947.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156947.hg.1 |
ENST00000564336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156947.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600156947.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156947.hg.1 |
ENST00000564336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156947.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600156947.hg.1 |
TC1600007819.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600156948.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156948.hg.1 |
ENST00000564336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156948.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600156948.hg.1 |
ENST00000268459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156948.hg.1 |
ENST00000564336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156948.hg.1 |
NM_033119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600156948.hg.1 |
TC1600007819.hg.1 |
Clariom D TC |