Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007674.hg.1 |
TC02000501.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007674.hg.1 |
ENST00000409785.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007674.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007680.hg.1 |
TC02000501.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007680.hg.1 |
ENST00000409785.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007680.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007681.hg.1 |
TC02000501.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007681.hg.1 |
ENST00000409785.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007681.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007682.hg.1 |
TC02000501.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007682.hg.1 |
ENST00000409785.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007682.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007675.hg.1 |
TC02000501.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007675.hg.1 |
ENST00000409785.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007675.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000428691.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000428691.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000428691.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000428691.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000453448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000453448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000453448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000453448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000453448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000453448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000456682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000456682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000456682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
ENST00000456682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007673.hg.1 |
TC02000501.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007673.hg.1 |
ENST00000409785.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007673.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156989.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156989.hg.1 |
ENST00000428691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156989.hg.1 |
ENST00000453448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156989.hg.1 |
ENST00000456682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156989.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156989.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156989.hg.1 |
ENST00000428691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156989.hg.1 |
ENST00000453448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156989.hg.1 |
ENST00000456682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156989.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156989.hg.1 |
TC0200008233.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200156979.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156979.hg.1 |
ENST00000456682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156979.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156979.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156979.hg.1 |
ENST00000456682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156979.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156979.hg.1 |
TC0200008233.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000428691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000428691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000428691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000428691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000453448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000453448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000453448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000453448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000453448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000453448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000456682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000456682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000456682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
ENST00000456682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008233.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156991.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156991.hg.1 |
ENST00000428691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156991.hg.1 |
ENST00000453448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156991.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156991.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156991.hg.1 |
ENST00000428691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156991.hg.1 |
ENST00000453448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156991.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156991.hg.1 |
TC0200008233.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200156980.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156980.hg.1 |
ENST00000456682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156980.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156980.hg.1 |
ENST00000409785.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156980.hg.1 |
ENST00000456682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156980.hg.1 |
NM_020122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156980.hg.1 |
TC0200008233.hg.1 |
Clariom D TC |