Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000509486.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000509486.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000509486.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000509486.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000509486.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000509486.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000514923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000514923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000514923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000514923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
ENST00000514923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016500.hg.1 |
TC05001151.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016500.hg.1 |
ENST00000382496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016500.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016501.hg.1 |
TC05001151.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016501.hg.1 |
ENST00000382496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016501.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170623.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170623.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170623.hg.1 |
ENST00000514923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170623.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170623.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170623.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170623.hg.1 |
ENST00000514923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170623.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170623.hg.1 |
TC0500010043.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000509486.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000509486.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000509486.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000509486.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000509486.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000509486.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000514923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000514923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000514923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000514923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
ENST00000514923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010043.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170622.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170622.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170622.hg.1 |
ENST00000514923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170622.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170622.hg.1 |
ENST00000382496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170622.hg.1 |
ENST00000513968.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170622.hg.1 |
ENST00000514923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170622.hg.1 |
NM_003966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170622.hg.1 |
TC0500010043.hg.1 |
Clariom D TC |