Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015512.4 | NP_056327.4 | 36 - 37 | 6030 | 84 | |
XM_017006130.1 | XP_016861619.1 | 37 - 38 | 8917 | 84 | ||
GenBank mRNA | AB037831.3 | - | 36 - 37 | 6030 | 84 | |
AB290163.1 | - | 36 - 37 | 6030 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons |