Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020213.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020213.hg.1 |
ENST00000483923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020213.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020213.hg.1 |
TC10001577.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000408492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000462874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000462874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000462874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000464808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000464808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000464808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000494941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000494941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000494941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
ENST00000494941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011577.hg.1 |
NR_031619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178871.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178871.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178871.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178871.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178871.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178871.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178871.hg.1 |
TC1000011577.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000370575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000408492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000462874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000462874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000462874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000464808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000464808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000464808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000483923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000494941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000494941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000494941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
ENST00000494941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NM_032709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011577.hg.1 |
NR_031619.1 |
Refseq |