Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013807.hg.1 |
ENST00000376980.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013807.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013807.hg.1 |
TC10001097.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008211.hg.1 |
ENST00000549267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376946.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376946.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376946.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376946.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376946.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376946.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376946.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376946.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000447548.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000447548.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000447548.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000447548.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000476103.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000476103.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000476103.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000476103.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000476103.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000476103.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000483085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000483085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000483085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
ENST00000483085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008211.hg.1 |
ENST00000549267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168300.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168300.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168300.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168300.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168300.hg.1 |
TC1000010006.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376946.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376946.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376946.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376946.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376946.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376946.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376946.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376946.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000376980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000447548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000447548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000447548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000447548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000476103.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000476103.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000476103.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000476103.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000476103.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000476103.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000483085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000483085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000483085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
ENST00000483085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010006.hg.1 |
NM_022365.3 |
Refseq |