Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |