Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002759.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002759.hg.1 |
ENST00000382031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002759.hg.1 |
ENST00000399453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002759.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002759.hg.1 |
TC15000329.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149986.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149986.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149986.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149986.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149986.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149986.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149986.hg.1 |
TC1500007060.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007060.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149985.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149985.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149985.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149985.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149985.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149985.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149985.hg.1 |
TC1500007060.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500149984.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149984.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149984.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149984.hg.1 |
ENST00000300231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149984.hg.1 |
ENST00000382031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149984.hg.1 |
NM_002373.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149984.hg.1 |
TC1500007060.hg.1 |
Clariom D TC |