Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025753.hg.1 |
TC07003372.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025753.hg.1 |
ENST00000310300.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025753.hg.1 |
ENST00000422462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025753.hg.1 |
ENST00000497245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025753.hg.1 |
NM_002319.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025751.hg.1 |
TC07003372.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025751.hg.1 |
ENST00000310300.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025751.hg.1 |
ENST00000422462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025751.hg.1 |
ENST00000497245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025751.hg.1 |
NM_002319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201402.hg.1 |
ENST00000310300.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201402.hg.1 |
ENST00000467201.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201402.hg.1 |
ENST00000497245.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201402.hg.1 |
ENST00000498539.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201402.hg.1 |
ENST00000619071.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201402.hg.1 |
NM_001289934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201402.hg.1 |
NM_002319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201402.hg.1 |
ENST00000310300.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201402.hg.1 |
ENST00000467201.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201402.hg.1 |
ENST00000497245.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201402.hg.1 |
ENST00000498539.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201402.hg.1 |
ENST00000619071.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201402.hg.1 |
NM_001289934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201402.hg.1 |
NM_002319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201402.hg.1 |
TC0700013598.hg.1 |
Clariom D TC |