Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003611.hg.1 |
TC09000338.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003611.hg.1 |
ENST00000376896.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003611.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003610.hg.1 |
TC09000338.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003610.hg.1 |
ENST00000376896.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003610.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003614.hg.1 |
TC09000338.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003614.hg.1 |
ENST00000376896.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003614.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003614.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151350.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151350.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151350.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151350.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151350.hg.1 |
TC0900007593.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900151349.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151349.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151349.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151349.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151349.hg.1 |
TC0900007593.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900151353.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151353.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151353.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151353.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151353.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151353.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151353.hg.1 |
TC0900007593.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000376896.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
ENST00000396204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007593.hg.1 |
NM_006914.3 |
Refseq |