Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010621.hg.1 |
TC04000773.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010621.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010620.hg.1 |
TC04000773.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010620.hg.1 |
ENST00000240488.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010620.hg.1 |
ENST00000504860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010620.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010620.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010620.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000503967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000503967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000503967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000503967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000504860.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000504860.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000504860.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000504860.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000504860.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000504860.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000504860.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000508731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000508731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000508731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000508731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000508731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000578990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000622785.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000622785.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000622785.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000622785.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000622785.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000622785.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000622785.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000622785.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
ENST00000622785.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000503967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000503967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000503967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000503967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000504860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000504860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000504860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000504860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000504860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000504860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000504860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000508731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000508731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000508731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000508731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000508731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000578990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000622785.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000622785.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000622785.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000622785.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000622785.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000622785.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000622785.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000622785.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
ENST00000622785.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009037.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162000.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162000.hg.1 |
ENST00000504860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162000.hg.1 |
ENST00000508731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162000.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162000.hg.1 |
ENST00000622785.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162000.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162000.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162000.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162000.hg.1 |
ENST00000240488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162000.hg.1 |
ENST00000504860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162000.hg.1 |
ENST00000508731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162000.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162000.hg.1 |
ENST00000622785.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162000.hg.1 |
NM_001253861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162000.hg.1 |
NM_032117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162000.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162000.hg.1 |
TC0400009037.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400162004.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162004.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162004.hg.1 |
ENST00000509752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162004.hg.1 |
NR_045605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162004.hg.1 |
TC0400009037.hg.1 |
Clariom D TC |