Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012063.3 | NP_036193.2 | 14 - 15 | 1755 | 87 | |
XM_011520543.2 | XP_011518845.1 | 15 - 16 | 1766 | 87 | ||
XM_017018664.1 | XP_016874153.1 | 13 - 14 | 1472 | 87 | ||
GenBank mRNA | AK302565.1 | - | 14 - 15 | 1755 | 87 | |
BC024590.1 | - | 14 - 15 | 1625 | 3882922 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |