Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383781.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383786.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000383788.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000603808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000604401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000604401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000604401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000604401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000605797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000605797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000605797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000605797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000605797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000605797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000605797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000605797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000605797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000605797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
ENST00000605797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_005677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013947.hg.1 |
NM_080539.3 |
Refseq |