Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000595713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000595713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19020809.hg.1 |
ENST00000221347.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19020809.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19020809.hg.1 |
TC19001527.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000595713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000595713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
ENST00000620799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010676.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900188148.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188148.hg.1 |
NM_003890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900188148.hg.1 |
TC1900010676.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900188148.hg.1 |
ENST00000616721.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188148.hg.1 |
NM_003890.2 |
Refseq |