Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs01553418_m1
See other CELF1 GE Assays ›
Gene Symbol:
CELF1
Gene Name:
CUGBP, Elav-like family member 1
Gene Aliases:
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
Chromosome Location:
Chr.11: 47465937 - 47565569 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs01553418_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: CELF1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001025596.2
NP_001020767.1
6 - 7
692
65
NM_001172639.1
NP_001166110.1
9 - 10
1654
65
NM_001172640.1
NP_001166111.1
6 - 7
689
65
XM_011519848.2
XP_011518150.1
7 - 8
764
65
XM_011519850.2
XP_011518152.2
7 - 8
1003
65
XM_011519852.2
XP_011518154.1
9 - 10
1112
65
XM_011519854.1
XP_011518156.1
11 - 12
1220
65
XM_011519855.1
XP_011518157.1
7 - 8
941
65
XM_011519856.1
XP_011518158.1
8 - 9
1022
65
XM_011519857.1
XP_011518159.1
7 - 8
893
65
XM_011519858.1
XP_011518160.1
7 - 8
988
65
XM_017017101.1
XP_016872590.1
10 - 11
1104
65
XM_017017102.1
XP_016872591.1
9 - 10
1023
65
XM_017017103.1
XP_016872592.1
8 - 9
916
65
XM_017017104.1
XP_016872593.1
8 - 9
916
65
XM_017017105.1
XP_016872594.1
8 - 9
916
65
XM_017017106.1
XP_016872595.1
10 - 11
1104
65
XM_017017107.1
XP_016872596.1
8 - 9
916
65
XM_017017115.1
XP_016872604.1
9 - 10
944
65
XM_017017116.1
XP_016872605.1
8 - 9
979
65
XM_017017117.1
XP_016872606.1
7 - 8
740
65
XM_017017118.1
XP_016872607.1
7 - 8
862
65
XM_017017119.1
XP_016872608.1
9 - 10
1081
65
XM_017017120.1
XP_016872609.1
7 - 8
893
65
XM_017017121.1
XP_016872610.1
7 - 8
988
65
XM_017017122.1
XP_016872611.1
7 - 8
893
65
XM_017017123.1
XP_016872612.1
7 - 8
941
65
XM_017017124.1
XP_016872613.1
9 - 10
1035
65
XM_017017125.1
XP_016872614.1
7 - 8
764
65
XM_017017126.1
XP_016872615.1
7 - 8
893
65
XM_017017127.1
XP_016872616.1
7 - 8
917
65
XM_017017128.1
XP_016872617.1
7 - 8
941
65
XM_017017129.1
XP_016872618.1
8 - 9
939
65
XM_017017130.1
XP_016872619.1
7 - 8
893
65
XM_017017131.1
XP_016872620.1
7 - 8
917
65
XM_017017132.1
XP_016872621.1
7 - 8
941
65
GenBank mRNA
AB210019.1
-
9 - 10
1654
65
AF248648.1
-
6 - 7
700
65
AF267533.1
-
6 - 7
693
65
AJ007988.1
-
6 - 7
682
65
AK304430.1
-
6 - 7
632
65
AK307679.1
-
1 - 2
154
65
Back To Top
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact
Tech Support
if you need more information.
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene CELF1
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splice site selection
mRNA processing
germ cell development
spermatid development
embryo development
RNA interference
negative regulation of translation
positive regulation of multicellular organism growth
regulation of RNA splicing
nucleotide binding
translation repressor activity, nucleic acid binding
RNA binding
mRNA binding
protein binding
translation initiation factor binding
BRE binding
poly(A) RNA binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0