Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16015332.hg.1 |
ENST00000311303.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16015332.hg.1 |
ENST00000416054.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16015332.hg.1 |
ENST00000448542.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16015332.hg.1 |
ENST00000449939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16015332.hg.1 |
NM_033226.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16015332.hg.1 |
TC16001093.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600178613.hg.1 |
ENST00000311303.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178613.hg.1 |
ENST00000497206.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178613.hg.1 |
ENST00000529084.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178613.hg.1 |
ENST00000529504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178613.hg.1 |
ENST00000532355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178613.hg.1 |
ENST00000532494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178613.hg.1 |
ENST00000534418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178613.hg.1 |
NM_033226.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178613.hg.1 |
ENST00000311303.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178613.hg.1 |
ENST00000497206.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178613.hg.1 |
ENST00000529084.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178613.hg.1 |
ENST00000529504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178613.hg.1 |
ENST00000532355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178613.hg.1 |
ENST00000532494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178613.hg.1 |
ENST00000534418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178613.hg.1 |
NM_033226.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178613.hg.1 |
TC1600010215.hg.1 |
Clariom D TC |