Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031414.hg.1 |
TC03001723.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031414.hg.1 |
ENST00000251775.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031414.hg.1 |
ENST00000536067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031414.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000465505.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000465505.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000465505.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000465505.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000465505.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000465505.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000473417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000473417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000473417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000473417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000473417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000473417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000473417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000482965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000482965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000482965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000482965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
ENST00000482965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000465505.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000465505.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000465505.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000465505.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000465505.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000465505.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000473417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000473417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000473417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000473417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000473417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000473417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000473417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000482965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000482965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000482965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000482965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
ENST00000482965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012252.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191515.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191515.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191515.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191515.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191515.hg.1 |
ENST00000251775.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191515.hg.1 |
ENST00000471751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191515.hg.1 |
NM_003794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191515.hg.1 |
NR_073435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191515.hg.1 |
TC0300012252.hg.1 |
Clariom D TC |