Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035581.hg.1 |
TC01002320.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035581.hg.1 |
ENST00000264211.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035581.hg.1 |
ENST00000374935.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035581.hg.1 |
ENST00000374937.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035581.hg.1 |
ENST00000400415.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035581.hg.1 |
ENST00000400422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035581.hg.1 |
ENST00000536266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035581.hg.1 |
ENST00000537738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035581.hg.1 |
NM_001198801.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035581.hg.1 |
NM_001198802.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035581.hg.1 |
NM_003760.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197718.hg.1 |
ENST00000264211.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197718.hg.1 |
ENST00000374935.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197718.hg.1 |
ENST00000400422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197718.hg.1 |
ENST00000602326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197718.hg.1 |
NM_001198801.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197718.hg.1 |
NM_001198802.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197718.hg.1 |
NM_003760.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197718.hg.1 |
ENST00000264211.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197718.hg.1 |
ENST00000374935.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197718.hg.1 |
ENST00000400422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197718.hg.1 |
ENST00000602326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197718.hg.1 |
NM_001198801.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197718.hg.1 |
NM_001198802.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197718.hg.1 |
NM_003760.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197718.hg.1 |
TC0100013194.hg.1 |
Clariom D TC |