Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |