Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023285.hg.1 |
TC06001912.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023285.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023283.hg.1 |
TC06001912.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023283.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023283.hg.1 |
ENST00000543031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023283.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023280.hg.1 |
TC06001912.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023280.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023280.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023281.hg.1 |
TC06001912.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023281.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023281.hg.1 |
ENST00000541327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023281.hg.1 |
ENST00000543031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023281.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185645.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185645.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185645.hg.1 |
TC0600012434.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012434.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185641.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185641.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185641.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185641.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185641.hg.1 |
TC0600012434.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600185643.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185643.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185643.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185643.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185643.hg.1 |
TC0600012434.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600185642.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185642.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185642.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185642.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185642.hg.1 |
TC0600012434.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600185644.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185644.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185644.hg.1 |
ENST00000369705.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185644.hg.1 |
NM_002395.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185644.hg.1 |
TC0600012434.hg.1 |
Clariom D TC |