Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000471683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000471683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000471683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000471683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000471683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000495422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000495422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000495422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000495422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000495422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
ENST00000495422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013483.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037401.hg.1 |
TC01002427.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037401.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037401.hg.1 |
ENST00000373824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037401.hg.1 |
ENST00000471683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037401.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037401.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000471683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000471683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000471683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000471683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000471683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000495422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000495422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000495422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000495422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000495422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
ENST00000495422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013483.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200151.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200151.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200151.hg.1 |
ENST00000471683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200151.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200151.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200151.hg.1 |
ENST00000263974.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200151.hg.1 |
ENST00000373824.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200151.hg.1 |
ENST00000471683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200151.hg.1 |
NM_001135218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200151.hg.1 |
NM_005644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200151.hg.1 |
TC0100013483.hg.1 |
Clariom D TC |