Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017009091.1 | XP_016864580.1 | 4 - 5 | 589 | 66 | |
XM_017009092.1 | XP_016864581.1 | 7 - 8 | 1701 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |