Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185059.1 | NP_001171988.1 | 25 - 26 | 4237 | 78 | |
NM_001185074.1 | NP_001172003.1 | 18 - 19 | 3639 | 78 | ||
NM_024617.3 | NP_078893.2 | 25 - 26 | 4347 | 78 | ||
XM_005252207.3 | XP_005252264.1 | 25 - 26 | 4299 | 78 | ||
XM_006717283.1 | XP_006717346.1 | 25 - 26 | 4299 | 78 | ||
XM_011519011.2 | XP_011517313.1 | 25 - 26 | 4299 | 78 | ||
XM_011519012.2 | XP_011517314.1 | 25 - 26 | 4247 | 78 | ||
XM_011519013.2 | XP_011517315.1 | 25 - 26 | 4916 | 78 | ||
XM_011519014.2 | XP_011517316.1 | 25 - 26 | 4518 | 78 | ||
XM_011519015.1 | XP_011517317.1 | 25 - 26 | 4299 | 78 | ||
XM_011519016.1 | XP_011517318.1 | 25 - 26 | 4299 | 78 | ||
XM_017015130.1 | XP_016870619.1 | 25 - 26 | 4915 | 78 | ||
XM_017015131.1 | XP_016870620.1 | 25 - 26 | 4557 | 78 | ||
XM_017015132.1 | XP_016870621.1 | 26 - 27 | 4462 | 78 | ||
GenBank mRNA | AB051498.1 | - | 17 - 18 | 2885 | 78 | |
AK023471.1 | - | 10 - 11 | 933 | 78 | ||
AK055948.1 | - | 13 - 14 | 2197 | 78 | ||
AL832026.1 | - | 18 - 19 | 3603 | 78 | ||
AL832193.1 | - | 26 - 27 | 4389 | 78 | ||
AL832965.1 | - | 1 - 2 | 100 | 78 | ||
AL832988.1 | - | 12 - 13 | 1182 | 78 | ||
BC144523.1 | - | 23 - 24 | 4031 | 78 | ||
BX641077.1 | - | 24 - 25 | 5697 | 78 | ||
CR933643.1 | - | 25 - 26 | 4211 | 78 | ||
CR933644.1 | - | 25 - 26 | 4789 | 78 | ||
CR936608.1 | - | 25 - 26 | 4291 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |