Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003180.hg.1 |
TC05000262.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003180.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003180.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007549.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500150856.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500150856.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500150856.hg.1 |
ENST00000252744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500150856.hg.1 |
NM_020928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500150856.hg.1 |
TC0500007549.hg.1 |
Clariom D TC |