Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015205.hg.1 |
TC0X001117.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015205.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015205.hg.1 |
ENST00000374299.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015205.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015205.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009972.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168094.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168094.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168094.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168094.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168094.hg.1 |
ENST00000298085.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168094.hg.1 |
ENST00000374299.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168094.hg.1 |
NM_001048164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168094.hg.1 |
NM_032803.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168094.hg.1 |
TC0X00009972.hg.1 |
Clariom D TC |