Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006701.hg.1 |
TC02000439.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006701.hg.1 |
ENST00000389501.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006701.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000258100.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000258100.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000258100.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000258100.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000258100.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000258100.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000258100.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000258100.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000413491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000413491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000413491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000413491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000413491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000413491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000413491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000443900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000443900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000443900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000443900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000443900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000443900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000474652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000474652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000474652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000474652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000474652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000474652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000474652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000474652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000477038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000477038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000477038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000477038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000486518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000486518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000486518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000491204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000491204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000491204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000491204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000491204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000629411.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000629411.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
ENST00000629411.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000258100.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000258100.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000258100.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000258100.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000258100.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000258100.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000258100.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000258100.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000413491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000413491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000413491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000413491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000413491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000413491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000413491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000443900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000443900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000443900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000443900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000443900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000443900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000474652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000474652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000474652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000474652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000474652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000474652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000474652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000474652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000477038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000477038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000477038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000477038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000486518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000486518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000486518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000491204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000491204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000491204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000491204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000491204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000629411.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000629411.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
ENST00000629411.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008040.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155515.hg.1 |
ENST00000258100.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155515.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155515.hg.1 |
ENST00000477038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155515.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155515.hg.1 |
ENST00000258100.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155515.hg.1 |
ENST00000389501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155515.hg.1 |
ENST00000477038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155515.hg.1 |
NM_006062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155515.hg.1 |
TC0200008040.hg.1 |
Clariom D TC |