Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005179.hg.1 |
TC03000255.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005179.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005179.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005179.hg.1 |
ENST00000465202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005179.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005179.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005177.hg.1 |
TC03000255.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005177.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005177.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005177.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005177.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000400888.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000400888.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000400888.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000400888.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000400888.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000400888.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000400888.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000400888.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000421659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000421659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000421659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000421659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000465202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000465202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000465202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000465202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
ENST00000465202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007256.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005178.hg.1 |
TC03000255.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005178.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005178.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005178.hg.1 |
ENST00000465202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005178.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005178.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151398.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151398.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151398.hg.1 |
ENST00000465202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151398.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151398.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151398.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151398.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151398.hg.1 |
ENST00000465202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151398.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151398.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151398.hg.1 |
TC0300007256.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300151397.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151397.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151397.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151397.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151397.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151397.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151397.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151397.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151397.hg.1 |
TC0300007256.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300151399.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151399.hg.1 |
ENST00000421659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151399.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151399.hg.1 |
ENST00000465202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151399.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151399.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151399.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151399.hg.1 |
ENST00000421659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151399.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151399.hg.1 |
ENST00000465202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151399.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151399.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151399.hg.1 |
TC0300007256.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000292301.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000400888.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000400888.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000400888.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000400888.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000400888.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000400888.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000400888.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000400888.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000421659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000421659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000421659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000421659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000445132.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000465202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000465202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000465202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000465202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
ENST00000465202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007256.hg.1 |
NM_001123396.2 |
Refseq |