Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005954.hg.1 |
TC03000289.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005954.hg.1 |
ENST00000296449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005954.hg.1 |
ENST00000309062.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005954.hg.1 |
ENST00000438782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005954.hg.1 |
ENST00000452691.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005954.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005954.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000366429.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000366429.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000366429.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000366429.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000366429.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000366429.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000366429.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000493870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
ENST00000493870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300152347.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300152347.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300152347.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300152347.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300152347.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300152347.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300152347.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300152347.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300152347.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300152347.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300152347.hg.1 |
TC0300007368.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000296449.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000366429.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000366429.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000366429.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000366429.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000366429.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000366429.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000366429.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000438782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000452691.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000493870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
ENST00000493870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_001135197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007368.hg.1 |
NM_178173.3 |
Refseq |