Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1900152763.hg.1 |
ENST00000221671.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152763.hg.1 |
ENST00000593380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152763.hg.1 |
ENST00000594035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152763.hg.1 |
ENST00000601288.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152763.hg.1 |
NM_001288834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900152763.hg.1 |
NM_032207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900152763.hg.1 |
TC1900007279.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900152763.hg.1 |
ENST00000221671.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152763.hg.1 |
ENST00000593380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152763.hg.1 |
ENST00000594035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152763.hg.1 |
ENST00000601288.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152763.hg.1 |
NM_001288834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900152763.hg.1 |
NM_032207.3 |
Refseq |