Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
ENST00000344788.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
ENST00000344788.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
ENST00000344788.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
ENST00000344788.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
ENST00000344788.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
ENST00000393655.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
ENST00000393655.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
ENST00000393655.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
ENST00000393655.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
ENST00000393655.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
ENST00000393655.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
ENST00000393655.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
ENST00000393655.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
ENST00000393655.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
ENST00000393655.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
ENST00000489228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
ENST00000489228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
ENST00000489228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
NM_003221.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
NM_003221.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
NM_003221.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
NM_003221.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
NM_003221.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
NM_003221.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
NM_003221.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
NM_003221.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
NM_003221.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008231.hg.1 |
NM_003221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
ENST00000344788.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
ENST00000344788.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
ENST00000344788.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
ENST00000344788.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
ENST00000344788.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
ENST00000393655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
ENST00000393655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
ENST00000393655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
ENST00000393655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
ENST00000393655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
ENST00000393655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
ENST00000393655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
ENST00000393655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
ENST00000393655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
ENST00000393655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
ENST00000489228.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
ENST00000489228.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
ENST00000489228.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
NM_003221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
NM_003221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
NM_003221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
NM_003221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
NM_003221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
NM_003221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
NM_003221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
NM_003221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
NM_003221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008231.hg.1 |
NM_003221.3 |
Refseq |