Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000532896.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000532896.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000532896.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000532896.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000532896.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000532896.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000532896.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000534421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000534421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000534421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000534421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000599612.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000599612.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000599612.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000599612.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000599612.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000599612.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000599612.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
ENST00000599612.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
NM_032683.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
NM_032683.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
NM_032683.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
NM_032683.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
NM_032683.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
NM_032683.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007366.hg.1 |
NM_032683.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004713.hg.1 |
ENST00000247712.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004713.hg.1 |
ENST00000534474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004713.hg.1 |
NM_032683.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004713.hg.1 |
TC19000353.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004710.hg.1 |
ENST00000247712.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004710.hg.1 |
NM_032683.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004710.hg.1 |
TC19000353.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900154053.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900154053.hg.1 |
ENST00000599612.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900154053.hg.1 |
NM_032683.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900154053.hg.1 |
TC1900007366.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900154053.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900154053.hg.1 |
ENST00000599612.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900154053.hg.1 |
NM_032683.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000532896.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000532896.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000532896.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000532896.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000532896.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000532896.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000532896.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000534421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000534421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000534421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000534421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000599612.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000599612.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000599612.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000599612.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000599612.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000599612.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000599612.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
ENST00000599612.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
NM_032683.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
NM_032683.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
NM_032683.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
NM_032683.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
NM_032683.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
NM_032683.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007366.hg.1 |
NM_032683.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900154055.hg.1 |
ENST00000532896.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900154055.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900154055.hg.1 |
ENST00000599612.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900154055.hg.1 |
NM_032683.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900154055.hg.1 |
TC1900007366.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900154055.hg.1 |
ENST00000532896.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900154055.hg.1 |
ENST00000533807.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900154055.hg.1 |
ENST00000599612.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900154055.hg.1 |
NM_032683.2 |
Refseq |