Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017746.hg.1 |
TC05001268.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017746.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017746.hg.1 |
ENST00000546130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017746.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000503535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000503535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000503535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000503535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000505524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000505524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000505524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000505524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
ENST00000505524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173044.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173044.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173044.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173044.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173044.hg.1 |
TC0500010493.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000296603.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000503535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000503535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000503535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000503535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000505524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000505524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000505524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000505524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
ENST00000505524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010493.hg.1 |
NM_001007527.1 |
Refseq |