Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000299.hg.1 |
ENST00000284894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000299.hg.1 |
ENST00000400546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000299.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000299.hg.1 |
TC21000060.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000461281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000461281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000461281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000461281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000461281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000461281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000484983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000484983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000484983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000486367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000486367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000486367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000486367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
ENST00000486367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000297.hg.1 |
ENST00000284894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000297.hg.1 |
ENST00000400546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000297.hg.1 |
ENST00000535285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000297.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000297.hg.1 |
TC21000060.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000296.hg.1 |
ENST00000284894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000296.hg.1 |
ENST00000400546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000296.hg.1 |
ENST00000535285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000296.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000296.hg.1 |
TC21000060.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2100145413.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100145413.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100145413.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100145413.hg.1 |
TC2100006714.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100145413.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100145413.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100145413.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100145414.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100145414.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100145414.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100145414.hg.1 |
TC2100006714.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100145414.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100145414.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100145414.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000284894.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000400546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000461281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000461281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000461281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000461281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000461281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000461281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000484983.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000484983.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000484983.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000486367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000486367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000486367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000486367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
ENST00000486367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006714.hg.1 |
NM_004540.4 |
Refseq |