Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025145.hg.1 |
ENST00000301200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025145.hg.1 |
ENST00000472703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025145.hg.1 |
NM_145057.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025145.hg.1 |
TC19001849.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025144.hg.1 |
ENST00000301200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025144.hg.1 |
ENST00000472703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025144.hg.1 |
NM_145057.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025144.hg.1 |
TC19001849.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025147.hg.1 |
ENST00000301200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025147.hg.1 |
NM_145057.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025147.hg.1 |
TC19001849.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011423.hg.1 |
ENST00000301200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011423.hg.1 |
ENST00000301200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011423.hg.1 |
ENST00000301200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011423.hg.1 |
ENST00000301200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011423.hg.1 |
ENST00000301200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011423.hg.1 |
ENST00000472703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011423.hg.1 |
ENST00000472703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011423.hg.1 |
ENST00000472703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900196545.hg.1 |
ENST00000301200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900196545.hg.1 |
TC1900011423.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900196545.hg.1 |
ENST00000301200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011423.hg.1 |
ENST00000301200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011423.hg.1 |
ENST00000301200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011423.hg.1 |
ENST00000301200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011423.hg.1 |
ENST00000301200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011423.hg.1 |
ENST00000301200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011423.hg.1 |
ENST00000472703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011423.hg.1 |
ENST00000472703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011423.hg.1 |
ENST00000472703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900196543.hg.1 |
ENST00000301200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900196543.hg.1 |
ENST00000472703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900196543.hg.1 |
TC1900011423.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900196543.hg.1 |
ENST00000301200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900196543.hg.1 |
ENST00000472703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900196542.hg.1 |
ENST00000301200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900196542.hg.1 |
ENST00000472703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900196542.hg.1 |
TC1900011423.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900196542.hg.1 |
ENST00000301200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900196542.hg.1 |
ENST00000472703.1 |
Ensembl |