Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000395103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000395103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000395103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022800.hg.1 |
TC08001729.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022800.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022800.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182988.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182988.hg.1 |
ENST00000395103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182988.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182988.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182988.hg.1 |
ENST00000395103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182988.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182988.hg.1 |
TC0800012160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800182989.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182989.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182989.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182989.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182989.hg.1 |
TC0800012160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000395103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000395103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000395103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |