Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003061.2 | NP_003052.2 | 22 - 23 | 2617 | 82 | |
GenBank mRNA | AB011537.3 | - | 22 - 23 | 2617 | 82 | |
AB017167.1 | - | 22 - 23 | 2597 | 82 | ||
BC146761.1 | - | 22 - 23 | 2617 | 9007200 | 82 | |
BC146851.1 | - | 22 - 23 | 2530 | 9056779 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |