Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000462394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000462394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000462394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000470950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000470950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000470950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000470950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000470950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000474283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000474283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000474283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000474283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000474283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000484476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000484476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000484476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000484476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000484476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000485476.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000485476.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004930.hg.1 |
TC01000323.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004930.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004930.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004930.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000462394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000462394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000462394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000470950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000470950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000470950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000470950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000470950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000474283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000474283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000474283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000474283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000474283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000484476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000484476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000484476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000484476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000484476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000485476.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000485476.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007406.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100151604.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151604.hg.1 |
ENST00000462394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151604.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151604.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100151604.hg.1 |
ENST00000374338.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151604.hg.1 |
ENST00000462394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151604.hg.1 |
ENST00000488127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151604.hg.1 |
NM_015627.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100151604.hg.1 |
TC0100007406.hg.1 |
Clariom D TC |