Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEZ6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184775.1 | NP_001171704.1 | 13 - 14 | 2991 | 95 | |
GenBank mRNA | BC143854.1 | - | 13 - 14 | 2890 | 9052370 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |