Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000492.3 | NP_000483.3 | 16 - 17 | 2786 | 175 | |
XM_011515751.2 | XP_011514053.1 | 17 - 18 | 2759 | 175 | ||
XM_011515753.1 | XP_011514055.1 | 17 - 18 | 2876 | 175 | ||
XM_011515754.2 | XP_011514056.1 | 19 - 20 | 3851 | 175 | ||
XM_017011699.1 | XP_016867188.1 | 18 - 19 | 3597 | 175 | ||
GenBank mRNA | M28668.1 | - | 16 - 17 | 2786 | 175 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Homo sapiens Probe spans exons |