Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251905.1 | NP_001238834.1 | 6 - 7 | 679 | 129 | |
NM_018171.3 | NP_060641.2 | 6 - 7 | 636 | 129 | ||
XM_017019551.1 | XP_016875040.1 | 6 - 7 | 1041 | 129 | ||
XM_017019552.1 | XP_016875041.1 | 6 - 7 | 1751 | 129 | ||
XM_017019553.1 | XP_016875042.1 | 7 - 8 | 3349 | 129 | ||
XM_017019554.1 | XP_016875043.1 | 6 - 7 | 555 | 129 | ||
GenBank mRNA | AK001521.2 | - | 6 - 7 | 456 | 129 | |
AK290562.1 | - | 6 - 7 | 489 | 129 | ||
AK293783.1 | - | 6 - 7 | 428 | 129 | ||
AK296610.1 | - | 6 - 7 | 435 | 129 | ||
AK310377.1 | - | 6 - 7 | 424 | 129 | ||
AY113704.1 | - | 6 - 7 | 636 | 129 | ||
BC033731.1 | - | 6 - 7 | 540 | 5168590 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |