Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000375322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000375322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000375322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000375322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000375322.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000413724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000413724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000413724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000413724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000413724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000415139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000415139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000415139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000415139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000415139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000415139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000415139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000430603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000430603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000430603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000430603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000430603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002465.hg.1 |
ENST00000263056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002465.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002465.hg.1 |
ENST00000375328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002465.hg.1 |
ENST00000542547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002465.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002465.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002465.hg.1 |
TC10000211.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000375322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000375322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000375322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000375322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000375322.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000413724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000413724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000413724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000413724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000413724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000415139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000415139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000415139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000415139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000415139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000415139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000415139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000430603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000430603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000430603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000430603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000430603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007176.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000148887.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148887.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148887.hg.1 |
ENST00000413724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148887.hg.1 |
ENST00000415139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148887.hg.1 |
ENST00000430603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148887.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148887.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000148887.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000148887.hg.1 |
ENST00000263056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148887.hg.1 |
ENST00000375321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148887.hg.1 |
ENST00000413724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148887.hg.1 |
ENST00000415139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148887.hg.1 |
ENST00000430603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148887.hg.1 |
ENST00000542547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148887.hg.1 |
NM_001244134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000148887.hg.1 |
NM_005204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000148887.hg.1 |
TC1000007176.hg.1 |
Clariom D TC |