Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010354.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010354.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010354.hg.1 |
TC20000773.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010357.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010357.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010357.hg.1 |
TC20000773.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010355.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010355.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010355.hg.1 |
TC20000773.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010356.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010356.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010356.hg.1 |
TC20000773.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000161991.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161991.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000161991.hg.1 |
TC2000008894.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000161991.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161991.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000161990.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161990.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000161990.hg.1 |
TC2000008894.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000161990.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161990.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008894.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000161992.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161992.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000161992.hg.1 |
TC2000008894.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000161992.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161992.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000161993.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161993.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000161993.hg.1 |
TC2000008894.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000161993.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161993.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000161994.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161994.hg.1 |
NM_005225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000161994.hg.1 |
TC2000008894.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000161994.hg.1 |
ENST00000343380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161994.hg.1 |
NM_005225.2 |
Refseq |