Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007905.hg.1 |
TC20001740.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007905.hg.1 |
ENST00000339987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007905.hg.1 |
ENST00000360779.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007905.hg.1 |
ENST00000381808.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007905.hg.1 |
ENST00000381812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007905.hg.1 |
ENST00000467129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007905.hg.1 |
NM_001199784.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007905.hg.1 |
NM_015685.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007905.hg.1 |
NM_080489.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170781.hg.1 |
NR_037661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170781.hg.1 |
TC2000009966.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000170781.hg.1 |
NR_037661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170719.hg.1 |
ENST00000339987.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170719.hg.1 |
ENST00000360779.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170719.hg.1 |
ENST00000381808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170719.hg.1 |
ENST00000381812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170719.hg.1 |
ENST00000467129.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170719.hg.1 |
NM_001199784.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170719.hg.1 |
NM_015685.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170719.hg.1 |
NM_080489.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170719.hg.1 |
TC2000009964.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000170719.hg.1 |
ENST00000339987.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170719.hg.1 |
ENST00000360779.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170719.hg.1 |
ENST00000381808.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170719.hg.1 |
ENST00000381812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170719.hg.1 |
ENST00000467129.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170719.hg.1 |
NM_001199784.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170719.hg.1 |
NM_015685.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170719.hg.1 |
NM_080489.4 |
Refseq |