Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022150.hg.1 |
ENST00000369209.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022150.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022150.hg.1 |
TC10001681.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000468935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000468935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000468935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000468935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000468935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000480802.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000480802.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000480802.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000481291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000481291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
ENST00000481291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000181651.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000181651.hg.1 |
ENST00000480802.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000181651.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000181651.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000181651.hg.1 |
ENST00000480802.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000181651.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000181651.hg.1 |
TC1000011933.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000369209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000468935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000468935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000468935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000468935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000468935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000480802.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000480802.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000480802.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000481291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000481291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
ENST00000481291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011933.hg.1 |
NM_025015.2 |
Refseq |