Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166698.1 | NP_001160170.1 | 2 - 3 | 279 | 81 | |
XM_006717702.3 | XP_006717765.1 | 3 - 4 | 195 | 81 | ||
XM_006717703.3 | XP_006717766.1 | 3 - 4 | 377 | 81 | ||
XM_011539491.2 | XP_011537793.1 | 2 - 3 | 207 | 81 | ||
XM_017015888.1 | XP_016871377.1 | 3 - 4 | 377 | 81 | ||
XM_017015889.1 | XP_016871378.1 | 3 - 4 | 195 | 81 | ||
XM_017015893.1 | XP_016871382.1 | 3 - 4 | 377 | 81 | ||
GenBank mRNA | AK316542.1 | - | 2 - 3 | 275 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |