Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164586.1 | NP_001158058.1 | 21 - 22 | 10615 | 72 | |
XM_005245580.3 | XP_005245637.1 | 21 - 22 | 10585 | 72 | ||
XM_011510120.2 | XP_011508422.1 | 21 - 22 | 10707 | 72 | ||
XM_011510121.2 | XP_011508423.1 | 21 - 22 | 10692 | 72 | ||
XM_017002787.1 | XP_016858276.1 | 20 - 21 | 10387 | 72 | ||
GenBank mRNA | AY245430.2 | - | 10 - 11 | 2737 | 72 | |
BC033185.1 | - | 23 - 24 | 3194 | 72 | ||
BC131529.1 | - | 10 - 11 | 2318 | 40036345 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |