Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037594.hg.1 |
TC01002444.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037594.hg.1 |
ENST00000336798.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037594.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037594.hg.1 |
ENST00000471567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037594.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037599.hg.1 |
TC01002444.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037599.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037599.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037593.hg.1 |
TC01002444.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037593.hg.1 |
ENST00000336798.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037593.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037593.hg.1 |
ENST00000471567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037593.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000471567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000471567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000471567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000471567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000487984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000487984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000487984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
ENST00000487984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013545.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037591.hg.1 |
TC01002444.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037591.hg.1 |
ENST00000336798.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037591.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037591.hg.1 |
ENST00000471567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037591.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200503.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200503.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200503.hg.1 |
ENST00000471567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200503.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200503.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200503.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200503.hg.1 |
ENST00000471567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200503.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200503.hg.1 |
TC0100013545.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100200506.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200506.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200506.hg.1 |
ENST00000471567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200506.hg.1 |
ENST00000487984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200506.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200506.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200506.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200506.hg.1 |
ENST00000471567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200506.hg.1 |
ENST00000487984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200506.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200506.hg.1 |
TC0100013545.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100200511.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200511.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200511.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200511.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200511.hg.1 |
TC0100013545.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100200505.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200505.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200505.hg.1 |
ENST00000471567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200505.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200505.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200505.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200505.hg.1 |
ENST00000471567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200505.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200505.hg.1 |
TC0100013545.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100200512.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200512.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200512.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200512.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200512.hg.1 |
TC0100013545.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000336798.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000339394.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000471567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000471567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000471567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000471567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000487984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000487984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000487984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
ENST00000487984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
NM_014654.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013545.hg.1 |
NM_014654.3 |
Refseq |