Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011530336.2 | XP_011528638.1 | 15 - 16 | 2283 | 61 | |
XM_011530337.2 | XP_011528639.1 | 15 - 16 | 2283 | 61 | ||
XM_011530338.2 | XP_011528640.1 | 14 - 15 | 2189 | 61 | ||
XM_017028919.1 | XP_016884408.1 | 14 - 15 | 2189 | 61 | ||
GenBank mRNA | AK092684.1 | - | 3 - 4 | 752 | 61 | |
AK095916.1 | - | 4 - 5 | 525 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |