Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000373231.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000373231.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000373231.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000373231.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000373231.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000373231.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000373231.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000373231.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000373231.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000503878.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000503878.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000503878.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000503878.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000503878.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
ENST00000503878.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
NM_031460.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
NM_031460.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
NM_031460.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
NM_031460.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
NM_031460.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
NM_031460.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
NM_031460.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011738.hg.1 |
NM_031460.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020220.hg.1 |
TC06001697.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020220.hg.1 |
ENST00000373231.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020220.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020220.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020220.hg.1 |
NM_031460.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180518.hg.1 |
ENST00000373231.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180518.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180518.hg.1 |
ENST00000503878.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180518.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180518.hg.1 |
NM_031460.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180518.hg.1 |
ENST00000373231.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180518.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180518.hg.1 |
ENST00000503878.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180518.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180518.hg.1 |
NM_031460.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180518.hg.1 |
TC0600011738.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000373231.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000373231.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000373231.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000373231.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000373231.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000373231.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000373231.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000373231.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000373231.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000453413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000503878.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000503878.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000503878.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000503878.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000503878.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
ENST00000503878.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
NM_001135111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
NM_031460.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
NM_031460.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
NM_031460.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
NM_031460.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
NM_031460.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
NM_031460.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
NM_031460.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011738.hg.1 |
NM_031460.3 |
Refseq |