Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004643.hg.1 |
ENST00000261893.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004643.hg.1 |
ENST00000413507.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004643.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004643.hg.1 |
NM_171846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004643.hg.1 |
TC15000580.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000413507.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000413507.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000413507.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000413507.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000413507.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000413507.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000413507.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000413507.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000557972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000557972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000557972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000557972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000559782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000559782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
ENST00000559782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_001288585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_001288585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_001288585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_001288585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_001288585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_001288585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_001288585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_001288585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_171846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_171846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_171846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_171846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_171846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_171846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_171846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007513.hg.1 |
NM_171846.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153222.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153222.hg.1 |
ENST00000413507.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153222.hg.1 |
ENST00000557972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153222.hg.1 |
NM_001288585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153222.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153222.hg.1 |
NM_171846.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153222.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153222.hg.1 |
ENST00000413507.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153222.hg.1 |
ENST00000557972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153222.hg.1 |
NM_001288585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153222.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153222.hg.1 |
NM_171846.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153222.hg.1 |
TC1500007513.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000261893.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000413507.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000413507.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000413507.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000413507.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000413507.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000413507.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000413507.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000413507.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000557972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000557972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000557972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000557972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000559782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000559782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
ENST00000559782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_001288585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_001288585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_001288585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_001288585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_001288585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_001288585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_001288585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_001288585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_032857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_171846.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_171846.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_171846.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_171846.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_171846.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_171846.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_171846.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007513.hg.1 |
NM_171846.3 |
Refseq |